• USA Home
  • S4628 - Secobarbital-d5 solution

EMAIL THIS PAGE TO A FRIEND
S4628 Sigma

Secobarbital-d5 solution

100 μg/mL

Synonym: Secobarbital-(allyl-d5) solution

Purchase

Properties

InChI Key   DAOMPCXXNPYORA-UHFFFAOYSA-N
drug control   Home Office Schedule 2; stupéfiant; kontrollierte Droge in Deutschlandregulated under CDSA - not available from Sigma-Aldrich Canada
concentration   100 μg/mL
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2930 4.1(6.1) / PGII
WGK Germany 
1

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

S5147 Secobarbital-d5, analytical standard, 97 atom % D

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?