EMAIL THIS PAGE TO A FRIEND
S4633 Sigma

[Tyr1]-Somatostatin

≥97% (HPLC)

Synonym: GH

  • CAS Number 59481-23-1

  • Empirical Formula (Hill Notation) C82H108N18O20S2

  • Molecular Weight 1729.97

  •  MDL number MFCD00076768

  •  PubChem Substance ID 24899612

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Peptides and Proteins, Peptides for Cell Biology, Somatostatins More...
InChI Key   HTAVEOUQMYLKNB-UHFFFAOYSA-N
assay   ≥97% (HPLC)
storage temp.   −20°C
Gene Information   human ... SST(6750)

Description

Amino Acid Sequence

Tyr-Gly-Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr-Phe-Thr-Ser-Cys [Disulfide Bridge: 3-14]

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?