• USA Home
  • S4763 - Sarcosinamide hydrochloride

EMAIL THIS PAGE TO A FRIEND
S4763 Sigma

Sarcosinamide hydrochloride

  • CAS Number 5325-64-4

  • Empirical Formula (Hill Notation) C3H8N2O · HCl

  • Molecular Weight 124.57

  •  MDL number MFCD00058283

  •  PubChem Substance ID 24899618

Purchase

Properties

InChI Key   VVIXOTCTYAILNP-UHFFFAOYSA-N
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?