EMAIL THIS PAGE TO A FRIEND
S4890 Sigma

SP-Trisacryl® M

aqueous ethanol suspension, 25-70 μm

Purchase

Properties

Related Categories Cation Exchange Media, Ion Exchange Chromatography, Molecular Biology, Protein Chromatography, Proteomics More...
form   aqueous ethanol suspension
particle size   25-70 μm
capacity   ≥150 meq/L
storage temp.   2-8°C

Description

Application

Strongly acidic cation exchanger

Features and Benefits

The matrix, poly(N-tris[hydroxymethyl]methyl acrylamide), offers the same advantages as SP-Trisacryl Plus except that it has somewhat less resistance to strongly alkaline conditions (recommended for use and storage between pH 1 and 11).

Physical form

Suspension in 1 M NaCl containing 20% ethanol

Legal Information

Trisacryl is a registered trademark of Pall Corporation

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1993 / PGIII
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?