• USA Home
  • S4933 - Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
S4933 Sigma

Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous glycerol solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, ST-ST
biological source   rabbit
antibody form   IgG fraction of antiserum
clone   polyclonal
form   buffered aqueous glycerol solution
mol wt   antigen mol wt 89 kDa
species reactivity   human, mouse, rat
application(s)   western blot (chemiluminescent): 1:1,000 using an extract of CNTF-treated human SK-N-MC cells
conjugate   unconjugated
shipped in   wet ice
storage temp.   −20°C

Description

Immunogen

synthetic peptide (CEADPGSAAP[pY]LKTK) corresponding to human STAT3, (amino acids 695-709) conjugated to BSA.

Specificity

The antibody recognizes tyrosine705 phosphorylated STAT3 protein.

Physical form

Solution in 0.01 M Tris, pH 7.4, containing 75 mM NaCl, 5 mg/ml bovine serum albumin, and 50% glycerol

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

comparable product

Product #

Description

Add to Cart

S2690 Anti-phospho-STAT3 (pTyr705) antibody produced in rabbit, affinity isolated antibody, buffered aqueous glycerol solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?