• USA Home
  • S5054 - Solvent–Detergent Removal Resin

EMAIL THIS PAGE TO A FRIEND
S5054 Sigma

Solvent–Detergent Removal Resin

saline suspension with ethanol, 50 μm mean particle size

Synonym: SDR HyperD® Resin

Purchase

Properties

Related Categories Affinity Chromatography, Molecular Biology, Protein Chromatography, Proteomics, Specialty Resins More...
form   saline suspension with ethanol
matrix   HyperD F
mean particle size   50 μm
storage temp.   2-8°C

Description

Application

Useful for the removal of detergents from protein samples.

Features and Benefits

Characteristics are tolerant of very high flow rates without bed compression, and high exchange capacity. The bead has an exclusion limit of 10,000 Da MW, and is highly efficient at removing biological detergents from samples. Stable to most common solvents, and pH′s from 2-12.

Other Notes

HyperD media are highly porous, rigid ceramic beads filled with a derivatized hydrogel.

Quantity

Binding capacity: 4-10 column volumes of sample can be treated, with up to 99.9% removal

Physical form

Suspension in 1 M NaCl with 20% ethanol.

Legal Information

HyperD is a registered trademark of Pall Corporation

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
UN 1170 3/PG 3
WGK Germany 
3
Flash Point(F) 
105 °F
Flash Point(C) 
40 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?