• USA Home
  • S5274 - Spacers for vertical standard gel, 16.5 cm wide
EMAIL THIS PAGE TO A FRIEND
S5274 Sigma
Spacers for vertical standard gel, 16.5 cm wide
size 0.75 mm × 28 cm
Purchase
Description
Packaging
Set includes two spacers. Rounded corner mates with gasket and bottom corners of rectangular glass plate. Gasket design eliminates the need for a bottom spacer.
Price and Availability
Detect Your Unseen Protein
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?