• USA Home
  • S5383 - Sphingomyelinase from human placenta

EMAIL THIS PAGE TO A FRIEND
S5383 Sigma

Sphingomyelinase from human placenta

buffered aqueous glycerol solution, ≥100 units/mg protein (Lowry)

Synonym: Sphingomyelin choline phosphohydrolase, Sphingomyelin phosphodiesterase

Purchase

Properties

Related Categories 3.1.x.x Acting on esters, 3.x.x.x Hydrolases, Biochemicals and Reagents, Cell Biology, Enzyme Class Index,
form   buffered aqueous glycerol solution
foreign activity   Phospholipase C ≤0.1%
storage temp.   −20°C
Gene Information   human ... SMPD1(6609), SMPD2(6610)

Description

Unit Definition

One unit will hydrolyze 1.0 nanomole of sphingomyelin to N-acylsphingosine and choline phosphate per hr at pH 5.0 at 37 °C.

Physical form

Solution in 50% glycerol, 25 mM potassium phosphate, pH 4.5, containing 0.1% TRITON® X-100 and 0.05 mM PMSF

Biochem/physiol Actions

Initiates the formation of sphingomyelin-based second messengers. Activates MAPK (mitogen-activated protein kinase) and SAPKs (stress-activated protein kinases); generates ceramide from sphingomyelin.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?