• USA Home
  • S5389 - Superoxide Dismutase from Escherichia coli

EMAIL THIS PAGE TO A FRIEND
S5389 Sigma

Superoxide Dismutase from Escherichia coli

lyophilized powder, ≥1,000 units/mg protein

Synonym: SOD, Superoxide: superoxide oxidoreductase

Purchase

Description

Analysis Note

For assay method, see McCord, J.M. and Fridovich,I., J. Biol. Chem., 244, 6049 (1969).

Biochem/physiol Actions

Catalyzes the dismutation of superoxide radicals to hydrogen peroxide and molecular oxygen. Plays a critical role in the defense of cells against the toxic effects of oxygen radicals. Competes with nitric oxide (NO) for superoxide anion (which reacts with NO to form peroxynitrite), thereby SOD promotes the activity of NO. SOD has also been shown to suppress apoptosis in cultured rat ovarian follicles, neural cell lines, and transgenic mice.

General description

Superoxide dismutases are a group of low molecular weight metalloproteins present in all aerobic cells of plants, animals and micro-organisms. They provide protection against damaging reactions with the superoxide radical anion (O2-) by catalyzing its disproportionation into oxygen and hydrogen peroxide.

Other Notes

Iron-containing enzyme

Unit Definition

One unit will inhibit reduction of cytochrome c by 50% in a coupled system with xanthine oxidase at pH 7.8 at 25 °C in a 3.0 mL reaction volume. Xanthine oxidase concentration should produce an initial ΔA550 of 0.025 ± 0.005 per min.

Physical form

Lyophilized powder containing Tris buffer salts

Application

The Cu,Zn superoxide dismutase from Escherichia coli retains monomeric structure at high protein concentration, which could be applied towards future assessments of altered subunit interaction in all the bacteriocupreins. Superoxide dismutase from Escherichia coli has also been used in a study to investigate the superoxide anion participattion in 2-oxoglutarate-dependent hydroxylation.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Protocols

Enzymatic Assay of Superoxide Dismutase

This procedure applies to all products that have a specification for superoxide dismutase activity by enzymatic determination.
Keywords: Catalysis, Reductions

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?