• USA Home
  • S5437 - Anti-SynGap antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
S5437 Sigma

Anti-SynGap antibody produced in rabbit

1 mg/mL, affinity isolated antibody, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Neurobiology, Antibodies to G Proteins and Cyclic Nucleotides,
species reactivity   rat, mouse
application(s)   western blot: suitable
clone   polyclonal
concentration   1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   mouse ... Syngap1(240057)
rat ... Syngap1(192117)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

GST fusion of amino acids 947-1167 of SynGap from rat

Physical form

Solution in phosphate buffered saline containing 0.05% sodium azide and 1 mg/ml bovine serum albumin.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?