• USA Home
  • S5532 - Storage container, lead acrylic for γ-emitters

EMAIL THIS PAGE TO A FRIEND
S5532 Sigma

Storage container, lead acrylic for γ-emitters

0.5 mm lead equivalent

Purchase

Properties

inside size   10 in. (25 cm) × 6 1/2 in. (16.5 cm) × 4 in. (10 cm)
thickness   1/2 in.

Description

Compatibility

For use with 125I and other low energy γ-emitters.

Other Notes

Note: Do not use with 32P.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?