• USA Home
  • S5563 - HIS-Select® High Capacity (HC) Nickel coated plates

EMAIL THIS PAGE TO A FRIEND
S5563 Sigma

HIS-Select® High Capacity (HC) Nickel coated plates

96-well, clear, polystyrene

Purchase

Properties

Related Categories HIS-Select, HIS-Select Affinity Purification, HIS-Select Plates, High Throughput Screening, Molecular Biology,
material   polystyrene
description   flat bottom
grade   (Protein purification of greater than 90%)
shelf life   Once opened, product to be used within one week.
capacity   ≥4 μg/well binding capacity
storage temp.   2-8°C
  2-8°C

Description

Application

Plates suitable for use with direct quantification (BCA, Bradford or Coomassie blue), western blotting,affinity purification and mass spectrometry.

General description

HIS-Select High Capacity (HC) Coated Plates are coated with a proprietary high-density nickel chelate matrix. This high capacity coating provides a greater per-well binding capacity than any other commercial histidine binding plate and can be used in a variety of applications including in-well protein quantification, high throughput purification, and post-capture analysis such as MALDI. These plates are also pre-treated to reduce non-specific binding. Manufactured under ISO 9002 in Sigma′s GMP facility.

HIS-Select HC 96 well plats are coated with a high-density nickel chelate matrix. This enables high capacity affinity binding of recombinant fusion protein with histidine tags.

Legal Information

HIS-Select is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?