• USA Home
  • S5644 - Saccharolactone-Agarose

EMAIL THIS PAGE TO A FRIEND
S5644 Sigma

Saccharolactone-Agarose

saline suspension

Purchase

Properties

Related Categories Affinity Chromatography, Molecular Biology, Protein Chromatography, Proteomics, Specialty Resins More...
form   saline suspension
matrix   Cross-linked 4% beaded agarose
matrix activation   cyanogen bromide
matrix attachment   carboxyl
matrix spacer   9 atoms
capacity   ~10 mg/mL binding capacity (β-glucuronidase)
storage temp.   2-8°C

Description

Physical form

Suspension in 0.5 M NaCl containing 0.02% thimerosal

Application

Saccharolactone-agarose is used in affinity chromatography, protein chromatography and dye resins. Saccharolactone-agarose has been used to purify and characterize β-glucuronidase from bull seminal plasma and to study its role in fertilization.

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?