• USA Home
  • S5688 - HIS-Select® High Sensitivity (HS) Nickel Coated Plates

EMAIL THIS PAGE TO A FRIEND
S5688 Sigma

HIS-Select® High Sensitivity (HS) Nickel Coated Plates

96-well, clear strip-well plate

Purchase

Properties

Related Categories HIS-Select, HIS-Select Plates, Molecular Biology, Proteomics, Purification and Detection,
material   polystyrene
description   flat bottom
shelf life   Unopened product is stable for 2 years. Opened product to be used within a week.
storage temp.   2-8°C

Description

Application

Captured HIS-tagged proteins can be detected by standard ELISA methods.

General description

HIS-Select Nickel coated High Sensitivity multiwell plates are manufactured by utilizing a unique non-charged hydrophilic linkage for the nickel-chelate. This proprietary coating technology reduces non-specific binding of recombinant fusion proteins as detected with conventional nickel chelate resins. HIS-Select Nickel coated high sensitivity coated plates are designed for low-level detection of recombinant fusion proteins with histidine-containing tags. Captured HIS-tagged proteins can be detected by standard ELISA methods. Manufactured under ISO 9002 in Sigma′s GMP facility, HIS-Select coated High Sensitivity plates can detect ≤1 ng/well of tagged fusion protein.

Legal Information

Covered by U.S. Patent No 6,623,655.

HIS-Select is a registered trademark of Sigma-Aldrich Co. LLC

Price and Availability


Amplified Detection
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?