EMAIL THIS PAGE TO A FRIEND
S5779 Sigma

Sodium selenite

plant cell culture tested, ~98%

Purchase

Properties

InChI Key   BVTBRVFYZUCAKH-UHFFFAOYSA-L
assay   ~98%
mp   >350 °C(lit.)
suitability   plant cell culture tested

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Supplemental Hazard Statements 
Contact with acids liberates toxic gas.
RIDADR 
UN2630 - class 6.1 - PG 1 - EHS - Selenites, HI: all (not BR)
WGK Germany 
2
RTECS 
VS7350000

Documents

Certificate of Analysis

Protocols & Articles

Articles

Dietary Antioxidants

Antioxidants protect biological systems from oxidative damage produced by oxygen-containing free radicals and from redoxactive transition metal ions such as iron, copper, and cadmium.1 During the oxi...
Biofiles Volume 2 Article 2
Keywords: Adhesion, Anti-inflammatory agents, Applications, Cancer, Cardiovascular, Catalysis, Diabetes, Diseases, Gene expression, Metabolism, Neurodegenerative Diseases, Peroxidations, Phosphorylations, Prebiotics, Probiotics, Reductions, Transcription, Transduction, Vitamins

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?