• USA Home
  • S5895 - Sangivamycin hydrate from Streptomyces rimosus

EMAIL THIS PAGE TO A FRIEND
S5895 Sigma

Sangivamycin hydrate from Streptomyces rimosus

Synonym: 7-Deaza-7-carbamoyladenosine hydrate

  • CAS Number 18417-89-5

  • Empirical Formula (Hill Notation) C12H15N5O5 · xH2O

  • Molecular Weight 309.28 (anhydrous basis)

  •  MDL number MFCD00214391

  •  PubChem Substance ID 24899678

Purchase

Description

Packaging

1, 10 mg in glass bottle

Application

Sangivamycin hydrate from Streptomyces rimosus may be used in PKC-mediated cell signaling studies.

Biochem/physiol Actions

A cytotoxic purine nucleoside, active against human cytomegalovirus (HCMV); inhibits protein kinase C.

Sangivamycin is a deazapurine nucleoside analog produced by Streptomyces rimosus that arrests cell cycle at the G2 phase. It inhibits the activity of protein kinase C (PKC) and rhodopsin kinase and at higher concentrations it also inhibits protein kinase A and b-adrenergic receptor kinase. The phosphorylation of histone H1 protein is also one mechanism by which Sangivamycin prevents the cell growth and acts as cytotoxic agent against a variety of human cancers.4,5,6

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGII
WGK Germany 
3
RTECS 
UY9355000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?