• USA Home
  • S6063 - Greiner Multiwell Strips

EMAIL THIS PAGE TO A FRIEND
S6063 Sigma

Greiner Multiwell Strips

high binding, wells flat bottom (1 × 12 well)

Synonym: Greiner Multiwell Strips

Purchase

Properties

Related Categories 96 Well Plates, Cell Culture, Drug Discovery, Greiner, Greiner 96-Well Multiwell Plates,
material   polystyrene plate
  wells flat bottom (1 × 12 well)
sterility   non-sterile
feature   lid: No
mfr. no.   Greiner 701061
maximum volume   388 μL
size   12 wells
working volume   350 μL
color   none
matrix active group   High binding surface treatment

Description

General description

Both medium-binding and high binding are offered. High binding plates and strips are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding are suggested.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?