• USA Home
  • S6067 - SYPRO® photographic filter
EMAIL THIS PAGE TO A FRIEND
S6067 Sigma
SYPRO® photographic filter
Purchase
Description
Caution
Do not freeze.
General description
Can be used to photograph gels or blots stained with any of the SYPRO protein stains: SYPRO Orange, SYPRO Red, SYPRO Ruby or SYPRO Tangerine protein gel stains, or SYPRO Rose Plus or SYPRO Ruby protein blot stains.
Please note that gelatin filters may not be suitable for all gel documentation and CCD instruments. Confirm suitability with the instrument manufacturer before using to avoid filter damage.
Legal Information
SYPRO is a registered trademark of Life Technologies
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?