EMAIL THIS PAGE TO A FRIEND
S6126 Sigma

Stigmasterol

~90%

Synonym: 3β-Hydroxy-24-ethyl-5,22-cholestadiene, 5,22-Stigmastadien-3β-ol

Purchase

Properties

assay   ~90%

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?