• USA Home
  • S6183 - Anti-STAT5B antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
S6183 Sigma

Anti-STAT5B antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies against Proteins/Bioactives/Markers/Receptors for Stem Cell Biology, Antibodies for Cell Biology, Antibodies for Epigenetics,
species reactivity   mouse, human
application(s)   immunoprecipitation: 4 μg using STAT5B from human Raji or A431 cell lysates
  western blot: 0.5-2 μg/mL using human A431 and mouse 3T3/A31 cell lysates
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt 90-92 kDa (doublet)
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... STAT5B(6777)
mouse ... Stat5b(20851)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to human STAT5B, amino acids 774-787, conjugated to KLH.

Physical form

Solution in 0.1 M Tris-glycine, pH 7.4, containing 0.15 M NaCl, and 0.05% sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence

All labs need water
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
WGK Germany  nwg
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?