| Related Categories | Labels and Markers, Labware, Markers More... |
| color | blue |
Writes on cold, wet surfaces. Quick-drying. Alcohol based permanent marker removed by alcohol.
Customers Also Viewed
lab marker, for use on plastic embedding cassettes and frosted end of microscope slides, black, 5/cs