• USA Home
  • S6315 - Nunc® PCR amplification plates
EMAIL THIS PAGE TO A FRIEND
S6315 Sigma
Nunc® PCR amplification plates
96 well Silicon spacer plate, natural, 1/pk
Synonym: 96 multiwell plate, 96 well PCR plate, 96 well amplification plate, 96 well microplate, 96 well microtiter plate, 96 well plate
Purchase
Properties
Related Categories 96 Well Plates, Labware, Manufacturer Browser, Multiwell Plates, Nunc,
material   natural silicon
packaging   pack of 1
mfr. no.   Nunc 259684
Description
General description
Nunc® PCR Amplification Plates
• Uniform thin wall construction of 0.2 mL well offers excellent heat transfer with maximum product yield for processing multiple PCR samples
• Rigid plate design without skirt fits GeNunc tube trade and holder (Nunc PCR amplificaiton workstation; product no. P4366), allowing easy manipulation by automated handling equipment
• Conical wells of plate extend above the base to permit sealing with strip caps or Amplification Tape 96 to prevent well to well contamination
• Strip caps have dome shape to eliminate condensation and use of oil overlays
• Amplification Tape 96 is offered in single, precut sheets
• Silicone Spacer Plate ensures even heat transfer when Amplification Tape 96 is used
• Plate and tape are certified RNAse and DNAse free
• Compatible with 96 well format thermal cyclers that hold a V-shaped aluminum block for 96 0.2 mL tubes
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?