• USA Home
  • S6558 - Anti-Ste20/PAK, Subdomain VI (From Yeast) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
S6558 Sigma

Anti-Ste20/PAK, Subdomain VI (From Yeast) antibody produced in rabbit

affinity isolated antibody, buffered aqueous glycerol solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, ST-ST
biological source   rabbit
antibody form   affinity isolated antibody
clone   polyclonal
form   buffered aqueous glycerol solution
species reactivity   yeast, rat, rabbit
application(s)   immunoprecipitation: 4 μg using Ste20 isoforms from 0.5–1 mg of a rabbit brain cytosol.
  western blot (chemiluminescent): 2 μg/mL using a whole brain extract from a rat or rabbit
conjugate   unconjugated
shipped in   wet ice
storage temp.   −20°C

Description

Immunogen

synthetic peptide corresponding to the subdomain VI of S. cerevisiae Ste20 (amino acids 738-752) conjugated to KLH.

Specificity

By immunoblotting, the antibody recognizes recombinant yeast Ste20 protein, expressed in Escherichia. coli. It also reacts with a 65 kDa protein in rabbit and rat brain that corresponds to PAK. PAK is the Ste20-like kinase that interacts with the small G proteins Rac and Cdc42. The antibody cross-reacts with other proteins in rat tissue extracts that may be homologs of PAK.

Physical form

Solution in 0.07 M Tris-glycine, pH 7.0, containing 30% glycerol

Price and Availability


Detect Your Unseen Protein
Safety & Documentation

Safety Information

WGK Germany 
1
Protocols & Articles

Articles

Antibody Basics

Immunoglobulins (Igs) are produced by B lymphocytes and secreted into plasma. The Ig molecule in monomeric form is a glycoprotein with a molecular weight of approximately 150 kDa that is shaped more ...
Keywords: Affinity chromatography, Centrifugation, Chromatography, Digestions, Direct immunofluorescence, Gene expression, High performance liquid chromatography, Immunofluorescence, Ion Exchange, Microscopy, Precipitation, Purification, Rheumatology, Scanning electron microscopy

Related Content

Antibody Explorer - Over 80,000 Products

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

comparable product

Product #

Description

Add to Cart

P2979 Anti-PAK3 antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous glycerol solution

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?