• USA Home
  • S6561 - Anti-Sodium Channel, Brain Type II (α Subunit) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
S6561 Sigma

Anti-Sodium Channel, Brain Type II (α Subunit) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym: Anti-BII, Anti-SP11II

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Ion Channels, Antibodies to Sodium Channels,
species reactivity   rat, mouse
application(s)   immunohistochemistry (frozen sections): 1:75 using using frozen brain tissue
  immunoprecipitation: suitable
  western blot (chemiluminescent): 1:150-1:300
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
storage temp.   −20°C
Gene Information   mouse ... Scn2a1(110876)
rat ... Scn2a1(24766)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 467-485 of the α subunit of rat type II voltage-gated sodium channel (VGSC, SP11II) with additional N-terminal lysine and tyrosine, conjugated to KLH. The epitope corresponds to the sequence in the intracellular loop between the I and II domains of type II VGSC α subunit.

Physical form

Lyophilized at ~0.3 mg/ml from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin, 5% sucrose, and 0.025% sodium azide.

Price and Availability

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?