EMAIL THIS PAGE TO A FRIEND
S6648 Sigma

Sera

Purchase

Properties

shipped in   dry ice
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

B9433 Bovine Serum, Adult, USA origin, sterile-filtered, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?