• USA Home
  • S6677 - S-100B Protein from bovine brain

EMAIL THIS PAGE TO A FRIEND
S6677 Sigma

S-100B Protein from bovine brain

≥70% (HPLC), lyophilized powder

Synonym: S100 beta, S100-Beta, S100b

Purchase

Properties

Related Categories Actin/Actin-binding Proteins, Cell Biology, Cell Signaling and Neuroscience, Cytoskeleton and Extracellular Matrix, General Stem Cell Biology,
assay   ≥70% (HPLC)
form   lyophilized powder
composition   Protein, ~90% biuret
storage temp.   −20°C
Gene Information   cow ... S100B(282059)

Description

Biochem/physiol Actions

Inhibits microtubule assembly.

General description

Ca2+ binding proteins found in glial cell cytoplasm in two major dimeric forms, S-100A (α,β) and S-100B (β,β), and one minor form, S-100 (α,α).

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

92210 Timestrip Plus 0 °C
06693 Timestrip Plus -20 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?