• USA Home
  • S6688 - Greiner Multiwell Strips

EMAIL THIS PAGE TO A FRIEND
S6688 Sigma

Greiner Multiwell Strips

size 8 wells , strip, clear polystyrene wells round bottom (8 well), surface treatment medium binding, non-sterile

Synonym: Greiner Multiwell Strips

Purchase

Properties

Related Categories 96 Well Plates, Greiner, Greiner 96-Well Multiwell Plates, Greiner Multiwell Plates, Greiner Multiwell Strip Plates, Strips and Frames,
material   clear polystyrene wells round bottom (8 well)
sterility   non-sterile
mfr. no.   Greiner 767001
maximum volume   388 μL
size   8 wells , strip
working volume   350 μL
matrix active group   medium binding surface treatment

Description

General description

Both medium-binding and high binding are offered. High binding plates and strips are recommended for ELISAs. High binding denotes a binding capacity of 600 ng/cm2 and scatter of less than 10%. Optimal protein binding is variable and comparative tests between high and medium binding are suggested.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?