• USA Home
  • S7074 - Monoclonal Anti-SPASTIN antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
S7074 Sigma

Monoclonal Anti-SPASTIN antibody produced in mouse

~1.0 mg/mL, clone Sp 6C6, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-ADPSP, Anti-FSP2, Anti-KIAA1083, Anti-SPAST, Anti-SPG4

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Cell and Organelle Proteins, Antibodies to Cytoskeleton,
species reactivity   rat, human, mouse
application(s)   immunocytochemistry: suitable
  indirect ELISA: suitable
  western blot: 0.5-1 μg/mL using HeLa total cell extract
clone   Sp 6C6, monoclonal
concentration   ~1.0 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG2a
mol wt   antigen mol wt ~55 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... SPAST(6683)
mouse ... Spast(50850)
rat ... Spast(362700)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

recombinant human spastin.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Personal Protective Equipment  Eyeshields, Gloves, half-mask respirator (US), multi-purpose combination respirator cartridge (US)
RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?