• USA Home
  • S7144 - Sera, abnormal (deficient) human

EMAIL THIS PAGE TO A FRIEND
S7144 Sigma

Sera, abnormal (deficient) human

Triiodothyronine/thyroxine (T3/T4)-deficient

Purchase

Properties

Related Categories Cell Culture, Fetal Bovine Serum (FBS) and Other Animal Sera, Human Sera More...
description   essentially lipid-free, lyophilized from the indicated amount of serum.
composition   T3 level, <0.1 μg/L RIA
  T4 level, <4 μg/L RIA
storage temp.   2-8°C

Description

Application

Triiodothyronine/thyroxine deficient human serum may be useful in studies on the effects of thyroid hormones on cell signaling, differentiation and survival in cell culture.

Price and Availability


Cell Freezing

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?