• USA Home
  • S7394 - Sera, abnormal (deficient) human

EMAIL THIS PAGE TO A FRIEND
S7394 Sigma

Sera, abnormal (deficient) human

Progesterone-deficient

Purchase

Properties

Related Categories Cell Culture, Fetal Bovine Serum (FBS) and Other Animal Sera, Human Sera More...
description   essentially lipid-free, lyophilized from the indicated amount of serum.
composition   Progesterone level, <0.12 μg/L RIA
storage temp.   2-8°C

Price and Availability


Cell Freezing

All labs need water

Stericup and Steritop Filter Units
Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?