• USA Home
  • S7438 - Spectronic 10 Genesys Spectrophotometer
EMAIL THIS PAGE TO A FRIEND
S7438 Sigma
Spectronic 10 Genesys Spectrophotometer
Synonym: spectroscopy uv visible, ultraviolet spectrophotometer, uv vis spectroscopy
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Labware, Manufacturer Browser, Photometers,
AC input   100 - 240 V, 50 - 60 Hz
W × D × H   33 cm (13 in.) × 41 cm (16 in.) × 23.5 cm (9 in.)
Description
General description
Features a xenon flashlamp and dual detectors for an outstanding signal-to-noise ratio. The xenon lamp flashes only when needed during measurements resulting in extended lamp life and lower operating costs. Concentration measurements only require two additional steps: measuring and entering the concentration of the standard. Simple keyboard-based operation and "Softkeys," specific to each screen, simplify setting up the instrument with your laboratory′s test parameters. Cell changer buttons rotate samples into position.
Power Requirements 100-240 V, 50/60 Hz
Price and Availability
Just Prove it! Spectroquant Prove
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?