• USA Home
  • S7439 - L-Selectin/Fc Chimera from mouse

EMAIL THIS PAGE TO A FRIEND
S7439 Sigma

L-Selectin/Fc Chimera from mouse

>90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder

Synonym: CD62L, DREG, LAM-1, LECAM-1, Leu-8, Leukocyte Selectin, MEL-14 antigen, TQ1

Purchase

Description

Analysis Note

Measured by its ability to bind L-Selectin ligands on LS180 cells.

General description

Extracellular domain of mouse L-Selectin (amino acids 1-332) fused to the C-terminal 6X histidine-tagged Fc region of human IgG1.

Physical form

Lyophilized from 0.2 μm filtered solution in phosphate buffered saline.

Biochem/physiol Actions

Cell surface glycoprotein involved in the migration of lymphocytes and neutrophils. Human and mouse L-Selectin share 76% sequence identity.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?