EMAIL THIS PAGE TO A FRIEND
S7440 Sigma
SlideWrite plus
Version 6, CD-ROM
Purchase
Properties
Related Categories Computer Software, Data Analysis software, Labware More...
Description
General description
Technical graphing, curve-fitting and data analysis functions that can transform your data into technical graphs and charts that are impressive and memorable. A technical presentation program for development of publication-quality graphs, charts, and scientific figures. Along with full statistics, curve-fitting, and data analysis features, SlideWrite also boasts drawing capabilities, optional libraries of scientific clip art, and a wide range of device drivers for slidemakers, printers, plotters, and cameras. Exports TIFF, PCX, CGM, EPS, and HPGL files; imports Lotus, ASCII, and PCX files, among others.
System Requirements
Windows 95, 98, 2000 or NT 4 and approximately 5MB of free disk space to install
Legal Information
SlideWrite is a trademark of Advanced Graphics Software
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?