• USA Home
  • S7522 - Storage container, acrylic, for β-emitters
EMAIL THIS PAGE TO A FRIEND
S7522 Sigma
Storage container, acrylic, for β-emitters
size 8 3/4 in. × 2 5/8 in. × 3 1/2 in.
Purchase
Properties
Related Categories Labware, Radiation Safety, Radiation Safety Containers, Safety Equipment
size   8 3/4 in. × 2 5/8 in. × 3 1/2 in.
Description
General description
Four chambers in one unit, each chamber with its own hinged lid, allows removal of one isotope without exposure to others. Outer panels of 1/2 in. thick arylic provide shielding against most β-emitters including 32P.
Each chamber is 1.6 in. square × 2.25 in. high (4 × 5.7 cm). Removable inserts reduce the chambers to 1.12 in. (2.8 cm) square to hold smaller vials upright.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?