• USA Home
  • S7525 - Finnpipette® repeat dispenser
EMAIL THIS PAGE TO A FRIEND
S7525 Sigma
Finnpipette® repeat dispenser
Syringe, size 20-200 μL
Purchase
Properties
Related Categories Finn Pipettors, Labware, Liquid Handling Equipment, Pipettes, Pipettors and Tips, Pipettors More...
mfr. no.   Finn
size   20-200 μL
Description
General description
Adjustable-volume positive-displacement pipet permits highly repetitive dispensing with the greatest possible accuracy and precision. Ergonomic design reduces operator fatigue. The 50 mL reservoir/valve assembly is easily removable, allowing quick change-over when working with several reagents at once. Disposable syringe fits between pipetting unit and valve, and provides the positive displacement action. Get extra capacity for very large dispensing tasks by connecting the inlet tube to a larger vessel. Dispensers come with one syringe and one bottle/valve assembly. Extras available separately.
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
comparable product
Product #
Description
Add to Cart
Z376892 Nichiryo digital hand-held dispenser, 2.0-10.0 mL dispensing volume
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?