• USA Home
  • S7564 - Solvent Safe pipette tips

EMAIL THIS PAGE TO A FRIEND
S7564 Sigma

Solvent Safe pipette tips

capacity 0.5-20 μL, sterile

Synonym: carbon fiber pipette tips, solvent safe pipette tips

Purchase

Properties

Related Categories Labware, Liquid Handling Equipment, MBP (Molecular BioProducts), MBP Pipette Tips, Manufacturer Browser,
material   carbon fiber
sterility   sterile
packaging   pkg of 4 trays (96 tips/tray)
mfr. no.   MBP, 5449P
L   2 in. (50.7 mm)
capacity   0.5-20 μL

Description

Compatibility

Compatibility:
Eppendorf 4810 and compatible pipettors
Eppendorf Research/Reference 2-20 μL
Eppendorf Research Pro 4860 5-100 μL
Eppendorf Titerman 4908 5-50 μL
Pipetman P-20 2-20 μL
Rainin EDP-Series 25
Socorex/Wheaton 822 Series

General description

• Solvent Safe carbon filter pipette tips
• Filters block corrosive vapors created when pipetting volatile organic solvents such as TFA
• Safely and accurately pipette phenol/chloroform DNA extractions
• Perform peptide synthesis and HPLC analysis
• Excellent alternative to glass pipettes

Legal Information

Solvent Safe is a trademark of Molecular BioProducts, Inc.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?