• USA Home
  • S7684 - Corning® multi-position stirrer
EMAIL THIS PAGE TO A FRIEND
S7684 Sigma
Corning® multi-position stirrer
positions, 4, AC input 230 V AC (with 12V DC remote controller)
Purchase
Properties
Related Categories Labware, Magnetic Stirrers, Stirrers More...
feature   positions 4
mfr. no.   Corning, 440836
availability   not available in EU
Description
Compatibility
Accepts up to 1 liter flasks. Ideal for use in incubator or fume hood.
General description
Available in four-, five-, and nine-position models. The speed of all positions is set simultaneously with either an integral knob or (an option on four-position units only) remote controller.
Legal Information
Corning is a registered trademark of Corning, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?