• USA Home
  • S7759 - Sarcosine Oxidase from Corynebacterium sp.

EMAIL THIS PAGE TO A FRIEND
S7759 Sigma

Sarcosine Oxidase from Corynebacterium sp.

lyophilized powder, 5-10 units/mg protein

Synonym: Sarcosine: oxygen oxidoreductase (demethylating)

Purchase

Description

Unit Definition

One unit will form 1.0 μmole of formaldehyde from sarcosine per min at pH 8.3 at 37 °C.

Physical form

Lyophilized powder containing phosphate buffer and lactose

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?