• USA Home
  • S7935 - Secure-Seal imaging spacers
EMAIL THIS PAGE TO A FRIEND
S7935 Sigma
Secure-Seal imaging spacers
diam. 9 mm
Purchase
Description
General description
8 chambers per spacer
Secure Seal imaging chambers are ultra thin adhesive spacers that peel and stick to glass coverslips or microscope slides to confine specimens without compression. Layer multiple spacers to custom build chambers to any depth desired. For high-resolution microscopy, sandwich specimen and spacer between two No. 0 glass coverslips. Individual spacers are 0.12 mm thick.
Legal Information
SecureSeal is a trademark of Grace Bio-Labs, Inc.
Price and Availability
Tissue Diagnostics
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?