• USA Home
  • S8026 - Streptokinase from β-hemolytic Streptococcus (Lancefield Group C)

EMAIL THIS PAGE TO A FRIEND
S8026 Sigma

Streptokinase from β-hemolytic Streptococcus (Lancefield Group C)

buffered aqueous solution, ≥70,000 units/mg protein (E1%/280)

Synonym: Streptokinase from Streptococcus hemolyticus

Purchase

Description

Application

Streptokinase is commonly used as a thrombolytic agent in the therapy of ischemic stroke. This therapy carries the important risk of intracerebral hemorrhage. Streptokinase is also used in the treatment of complicated parapneumonic effusions and empyema where adverse reactions, allergic type, are rare.

Unit Definition

One unit will liquify a standard clot of fibrinogen, plasminogen, and thrombin at pH 7.5 at 37 °C in 10 min.

Physical form

Solution containing 60 mM phosphate buffer and 0.6 M glutamate

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles

Articles

Plasma Derived Proteins and Enzymes

Despite their complexity, blood and plasma are abundant biological resources for the discovery of drug targets and biomarkers for human disease. It is estimated that plasma may contain as many as 40,...
BioFiles 2006, 1.5, 2.
Keywords: Applications, Gas chromatography, Type

Related Content

Enzymes & Proteins

Application Index | Enzyme Index | Substrate Index | Inhibitor Index | Cofactor Index | Lectin Index
Keywords: Cell signaling, Diagnostic, Drug discovery, Molecular biology

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

replaced by

Product #

Description

Add to Cart

S3134 Streptokinase from β-hemolytic Streptococcus (Lancefield Group C), lyophilized powder, ≥3,500 units/mg solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?