• USA Home
  • S8189 - Solvent Safe pipette tips
EMAIL THIS PAGE TO A FRIEND
S8189 Sigma
Solvent Safe pipette tips
volume 100-1000 μL
Purchase
Properties
Related Categories Labware, Liquid Handling Equipment, MBP (Molecular BioProducts), MBP Pipette Tips, Manufacturer Browser,
sterility   non-sterile
packaging   pkg of 500 (bag)
mfr. no.   Molecular BioProducts
L   3.5 in. (88.3 mm)
volume   100-1000 μL
Description
General description
• Solvent Safe carbon filter pipette tips
• Filters block corrosive vapors created when pipetting volatile organic solvents such as TFA
• Safely and accurately pipette phenol/chloroform DNA extractions
• Perform peptide synthesis and HPLC analysis
• Excellent alternative to glass pipettes
Legal Information
Solvent Safe is a trademark of Molecular BioProducts, Inc.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?