• USA Home
  • S8262 - Sodium phosphate glass

EMAIL THIS PAGE TO A FRIEND
S8262 Sigma

Sodium phosphate glass

Type 75+

Purchase

Properties

mol wt   mol wt ≥75 P (av. chain length)
storage temp.   2-8°C

Description

Biochem/physiol Actions

Reported to be of use in protein binding and sequestering and in studies on bone calcification.

Other Notes

Nan+2PnO3n+1; n = the average number of phosphorus (P) atoms in the chain.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Related Products

comparable product

Product #

Description

Add to Cart

S6253 Sodium phosphate glass, Type 65

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?