• USA Home
  • S8385 - [pGlu5, N-Me-Phe8, Sar9]-Substance P Fragment 5-11

EMAIL THIS PAGE TO A FRIEND
S8385 Sigma

[pGlu5, N-Me-Phe8, Sar9]-Substance P Fragment 5-11

≥95% (HPLC)

Synonym: pGlu-Gln-Phe-N-Methyl-Phe-Sar-Leu-Met-NH2

Purchase

Properties

InChI Key   IEEBRBMEXFNVKU-UHFFFAOYSA-N
assay   ≥95% (HPLC)
storage temp.   −20°C
Gene Information   human ... TAC1(6863)

Description

Biochem/physiol Actions

Substance P analog which is resistant to attack by a substance P degrading enzyme.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?