• USA Home
  • S8526 - Sodium hydroxide standard solution

EMAIL THIS PAGE TO A FRIEND
S8526 Sigma

Sodium hydroxide standard solution

for electrophoresis, 1.0 M

Purchase

Properties

grade   for electrophoresis
InChI Key   HEMHJVSKTPXQMS-UHFFFAOYSA-M
concentration   1.0 M

Description

Application

1 M sodium hydroxide is suitable as a concentrated cathode solution for isoelectric focusing (IEF). A 0.1 M sodium hydroxide solution is commonly used as a cathode solution but some procedures use other concentrations. The 1 M sodium hydroxide solution may be diluted to the desired concentration with deionized water.

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 1824 8/PG 3
WGK Germany 
1
RTECS 
WB4900000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

S2770 Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?