• USA Home
  • S8558 - mouse anti-Sulfanilic acid, purified mouse immunoglobulin

EMAIL THIS PAGE TO A FRIEND
S8558 Sigma

mouse anti-Sulfanilic acid, purified mouse immunoglobulin

Purchase

Properties

storage temp.   −20°C

Description

Other Notes

custom antibody preparation S R A dept 96 need 1100, 1200, 2100, 8155 and 9930 quantity one lot add to select deck

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?