EMAIL THIS PAGE TO A FRIEND
S8879 Sigma

Salicylaldoxime

Synonym: 2-Hydroxybenzaldehyde oxime

  • CAS Number 94-67-7

  • Linear Formula 2-(HO)C6H4CH=NOH

  • Molecular Weight 137.14

  •  Beilstein Registry Number 1859881

  •  EC Number 202-353-8

  •  MDL number MFCD00002120

  • Popular Documents:  FTNMR (PDF)  

Purchase

Properties

InChI Key   ORIHZIZPTZTNCU-VMPITWQZSA-N
mp   59-61 °C(lit.)
storage temp.   2-8°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
RTECS 
VN5775000
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

84172 Salicylaldoxime, ≥98.0% (NT)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?