• USA Home
  • S8937 - Cameo Filtration Membrane
EMAIL THIS PAGE TO A FRIEND
S8937 Sigma
Cameo Filtration Membrane
Nylon membrane (with glass prefilter), pore size 0.22 μm, volume 12 mL
Purchase
Properties
Related Categories Filtration Supplies, Labware, Nylon Membrane Syringe Filters, Syringe Filters, Disposable
material   Nylon membrane (with glass prefilter)
sterility   non-sterile
mfr. no.   Cameo 17GN
volume   12 mL
pore size   0.22 μm
fitting   (Slip)
Description
General description
Cameo 17 mm and 30 mm syringe filters offer 20% greater effective surface area over smaller 13 mm and 25 mm sizes for improved performance in HPLC sample preparation. Designed to speed up and increase sample volume throughput while reducing back pressure, the hold-up volume is slightly lower due to improved flow channel design and reduced spacing between the supports. The design improvements allow an increase in operating pressure the syringe filters can withstand.
Legal Information
Cameo is a trademark of GVS S.p.A.
Price and Availability
Sterifil Aseptic System and Holder
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?