EMAIL THIS PAGE TO A FRIEND
S9126 Sigma

5-Sulfaminouracil

~95%

Synonym: 2,4-Dihydroxy-5-sulfaminopyrimidine

  • CAS Number 5435-16-5

  • Empirical Formula (Hill Notation) C4H5N3O5S

  • Molecular Weight 207.16

  •  MDL number MFCD00083080

Purchase

Properties

Related Categories Biochemicals and Reagents, Nucleoside Analogs, Nucleosides, Nucleotides, Oligonucleotides More...
InChI Key   ZZRMMIWUJLCXPF-UHFFFAOYSA-N
assay   ~95%
storage temp.   −20°C

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?