• USA Home
  • S9141 - Hamilton® GASTIGHT® 1700 series syringe
EMAIL THIS PAGE TO A FRIEND
S9141 Sigma
Hamilton® GASTIGHT® 1700 series syringe
Purchase
Description
General description
250μl capacity, Model 1725N
For gas and liquid chromatography, corrosive gases, and volatile liquids. Chemically inert PTFE-tipped plunger. Leaktight syringe with 200 psi pressure rating. Cemented and removable needle models have 2 in. L, point style 2 needles.
Needles are not included with the Luer tip and TLL syringes.
*Indicates plunger is PTFE-coated.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?