• USA Home
  • S9190 - SigmaScreen Streptavidin High Capacity Coated Plates
EMAIL THIS PAGE TO A FRIEND
S9190 Sigma
SigmaScreen Streptavidin High Capacity Coated Plates
384-well black clear bottom
Purchase
Properties
material   polystyrene
description   flat bottom
storage temp.   2-8°C
Description
General description
SigmaScreen Streptavidin coated high capacity plates utilize a proprietary coating technology which provides substantially greater biotin binding capacity than standard Streptavidin or ExtrAvidin coated plates. With greater than 300 pmol/well for the 96-well clear format and greater than 150 pmol/well for the 384-well clear format, Streptavidin coated high capacity plates provide a platform which allows the captured protein to be eluted for post-capture analysis by various methods such as MALDI, ICAT or SDS-PAGE. Streptavidin coated high capacity plates are manufactured under ISO 9002 in Sigma’s GMP facility and deliver high well-to-well and plate-to-plate consistency.
Legal Information
SigmaScreen is a trademark of Sigma-Aldrich Co. LLC
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?